Assay Details
| Gene Symbol: Usp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038589.2 | NP_001033678.1 | 9 - 10 | 993 | 137 | |
| NM_021522.4 | NP_067497.2 | 10 - 11 | 1098 | 137 | ||
| GenBank mRNA | AB034633.1 | - | 10 - 11 | 877 | 137 | |
| AK011322.1 | - | 10 - 11 | 1034 | 137 | ||
| AK029977.1 | - | 10 - 11 | 1027 | 137 | ||
| AK045909.1 | - | 10 - 11 | 1018 | 137 | ||
| AK170933.1 | - | 1092 | 137 | |||
| AK172477.1 | - | 10 - 11 | 1085 | 137 | ||
| BC005571.1 | - | 10 - 11 | 1017 | 3157723 | 137 | |
| BC050197.1 | - | 9 - 10 | 911 | 30006882 | 137 | |
| CK621440.1 | - | 4 - 5 | 319 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp14
Genomic Map