Assay Details
| Gene Symbol: Usp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038589.2 | NP_001033678.1 | 11 - 12 | 1145 | 125 | |
| NM_021522.4 | NP_067497.2 | 12 - 13 | 1250 | 125 | ||
| GenBank mRNA | AK011322.1 | - | 12 - 13 | 1186 | 125 | |
| AK029977.1 | - | 12 - 13 | 1179 | 125 | ||
| AK045909.1 | - | 12 - 13 | 1170 | 125 | ||
| AK170933.1 | - | 1244 | 125 | |||
| AK172477.1 | - | 12 - 13 | 1236 | 124 | ||
| BC005571.1 | - | 12 - 13 | 1169 | 3157723 | 125 | |
| BC050197.1 | - | 11 - 12 | 1063 | 30006882 | 125 | |
| CK621440.1 | - | 6 - 7 | 471 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp14
Genomic Map