Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000007602.mm.1 |
NM_001033375.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007602.mm.1 |
NM_001033375.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007602.mm.1 |
TC1000001010.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000007599.mm.1 |
NM_001033375.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007599.mm.1 |
NM_001033375.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007599.mm.1 |
TC1000001010.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000007601.mm.1 |
NM_001033375.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007601.mm.1 |
NM_001033375.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007601.mm.1 |
TC1000001010.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000007600.mm.1 |
NM_001033375.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007600.mm.1 |
NM_001033375.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007600.mm.1 |
TC1000001010.mm.1 |
MTA TC |