Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000007578.mm.1 |
NM_001033375.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007578.mm.1 |
NM_001033375.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007578.mm.1 |
TC1000001010.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000007580.mm.1 |
NM_001033375.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007580.mm.1 |
NM_001033375.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007580.mm.1 |
TC1000001010.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000007577.mm.1 |
NM_001033375.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007577.mm.1 |
NM_001033375.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007577.mm.1 |
TC1000001010.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000007576.mm.1 |
NM_001033375.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007576.mm.1 |
NM_001033375.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007576.mm.1 |
TC1000001010.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000007579.mm.1 |
NM_001033375.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007579.mm.1 |
NM_001033375.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007579.mm.1 |
TC1000001010.mm.1 |
MTA TC |