Assay Details
Gene Symbol: Appl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145220.2 | NP_660255.1 | 9 - 10 | 786 | 63 | |
GenBank mRNA | AK154525.1 | - | 9 - 10 | 810 | 63 | |
AK160075.1 | - | 9 - 10 | 765 | 63 | ||
AK160105.1 | - | 9 - 10 | 762 | 63 | ||
AK161003.1 | - | 9 - 10 | 793 | 63 | ||
AK168748.1 | - | 9 - 10 | 742 | 63 | ||
AK171512.1 | - | 9 - 10 | 742 | 63 | ||
AY113706.1 | - | 9 - 10 | 788 | 63 | ||
BC048906.1 | - | 9 - 10 | 724 | 6475576 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
