Assay Details
| Gene Symbol: Ugp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139297.6 | NP_647458.1 | 1 - 2 | 438 | 124 | |
| GenBank mRNA | AF424698.1 | - | 1 - 2 | 286 | 124 | |
| AK033445.1 | - | 1 - 2 | 285 | 124 | ||
| BC023810.1 | - | 1 - 2 | 289 | 5353993 | 124 | |
| BC061208.1 | - | 1 - 2 | 325 | 30250244 | 124 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF424698.1, AK033445.1, BC023810.1, BC061208.1, NM_139297.6 transcripts.
Set Membership: |
|

Ugp2
Genomic Map