Assay Details
| Gene Symbol: Ugp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290634.1 | NP_001277563.1 | 3 - 4 | 493 | 130 | |
| NM_139297.6 | NP_647458.1 | 3 - 4 | 683 | 130 | ||
| GenBank mRNA | AF424698.1 | - | 3 - 4 | 531 | 130 | |
| AK033445.1 | - | 3 - 4 | 530 | 130 | ||
| AK153885.1 | - | 2 - 3 | 1626 | 130 | ||
| AK214438.1 | - | 3 - 4 | 314 | 130 | ||
| BC023810.1 | - | 3 - 4 | 534 | 5353993 | 130 | |
| BC025585.1 | - | 3 - 4 | 438 | 5324928 | 130 | |
| BC026626.1 | - | 2 - 3 | 220 | 130 | ||
| BC061208.1 | - | 3 - 4 | 570 | 30250244 | 130 | |
| CJ098156.1 | - | 3 - 4 | 493 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ugp2
Genomic Map