Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100023581.mm.1 |
NM_013552.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023581.mm.1 |
NM_013552.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023581.mm.1 |
TC1100002549.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100023582.mm.1 |
NM_013552.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023582.mm.1 |
NM_013552.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023582.mm.1 |
TC1100002549.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100023584.mm.1 |
NM_013552.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023584.mm.1 |
NM_013552.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023584.mm.1 |
TC1100002549.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100023585.mm.1 |
NM_013552.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023585.mm.1 |
NM_013552.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023585.mm.1 |
TC1100002549.mm.1 |
MTA TC |