Assay Details
| Gene Symbol: Hmmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013552.2 | NP_038580.2 | 9 - 10 | 975 | 69 | |
| GenBank mRNA | AF031932.1 | - | 9 - 10 | 960 | 69 | |
| AF079222.1 | - | 10 - 11 | 924 | 69 | ||
| AK143123.1 | - | 9 - 10 | 975 | 69 | ||
| AK163410.1 | - | 9 - 10 | 918 | 69 | ||
| X64550.1 | - | 5 - 6 | 588 | 69 | ||
More Information
Set Membership: |
|

Hmmr
Genomic Map