Assay Details
| Gene Symbol: Cabp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144532.2 | NP_653115.1 | 2 - 3 | 442 | 65 | |
| GenBank mRNA | AK171863.1 | - | 6 - 7 | 1052 | 65 | |
| AY039218.1 | - | 2 - 3 | 437 | 65 | ||
| BC029002.1 | - | 2 - 3 | 350 | 65 | ||
| BC049263.1 | - | 2 - 3 | 442 | 5366999 | 65 | |
| BC053330.1 | - | 2 - 3 | 397 | 65 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cabp4
Genomic Map