Assay Details
| Gene Symbol: Mtl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286557.1 | NP_001273486.1 | 2 - 3 | 565 | 58 | |
| GenBank mRNA | AF329360.1 | - | 2 - 3 | 575 | 58 | |
| U67176.1 | - | 3 - 4 | 498 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF329360.1, NM_001286557.1, U67176.1 transcripts.
Set Membership: |
|

Mtl5
Genomic Map