Assay Details
| Gene Symbol: Doc2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021791.3 | NP_068563.2 | 7 - 8 | 774 | 65 | |
| GenBank mRNA | BC061104.1 | - | 7 - 8 | 774 | 30296398 | 65 |
More Information
Updated Mapping Information
Set Membership: |
|

Doc2g
Genomic Map