Assay Details
Gene Symbol: Sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110791.1 | NP_001104261.1 | 6 - 7 | 1057 | 106 | |
NM_011750.2 | NP_035880.2 | 6 - 7 | 1057 | 106 | ||
NR_122118.1 | 5 - 6 | 981 | 106 | |||
GenBank mRNA | AK133895.1 | - | 6 - 7 | 846 | 106 | |
AK146151.1 | - | 6 - 7 | 818 | 106 | ||
AK147081.1 | - | 6 - 7 | 826 | 106 | ||
AK152914.1 | - | 5 - 6 | 759 | 106 | ||
AK157845.1 | - | 6 - 7 | 826 | 106 | ||
BC009091.1 | - | 6 - 7 | 802 | 3157495 | 106 | |
BC055370.1 | - | 6 - 7 | 816 | 30055109 | 106 | |
X85802.1 | - | 6 - 7 | 814 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
