Assay Details
| Gene Symbol: Lonp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028782.2 | NP_083058.2 | 12 - 13 | 1896 | 73 | |
| GenBank mRNA | AK004820.1 | - | 12 - 13 | 1880 | 73 | |
| AK131910.1 | - | 12 - 13 | 1882 | 73 | ||
| AK159302.1 | - | 12 - 13 | 1881 | 73 | ||
| AK160071.1 | - | 12 - 13 | 1881 | 73 | ||
| AK160076.1 | - | 12 - 13 | 1882 | 73 | ||
| AK161983.1 | - | 12 - 13 | 1881 | 73 | ||
| AY190302.1 | - | 12 - 13 | 1866 | 73 | ||
| BC057365.1 | - | 10 - 11 | 3290 | 73 | ||
| BC058735.1 | - | 10 - 11 | 3290 | 73 | ||
| BC092212.1 | - | 3 - 4 | 263 | 73 | ||
More Information
Set Membership: |
|

Lonp1
Genomic Map