Assay Details
| Gene Symbol: Lonp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028782.2 | NP_083058.2 | 13 - 14 | 2016 | 84 | |
| GenBank mRNA | AK004820.1 | - | 13 - 14 | 2000 | 84 | |
| AK131910.1 | - | 13 - 14 | 2002 | 84 | ||
| AK157474.1 | - | 13 - 14 | 1995 | 84 | ||
| AK159302.1 | - | 13 - 14 | 2001 | 84 | ||
| AK160071.1 | - | 13 - 14 | 2001 | 84 | ||
| AK160076.1 | - | 13 - 14 | 2002 | 84 | ||
| AK161983.1 | - | 13 - 14 | 2001 | 84 | ||
| AY190302.1 | - | 13 - 14 | 1986 | 84 | ||
| BC057365.1 | - | 11 - 12 | 3410 | 84 | ||
| BC058735.1 | - | 11 - 12 | 3410 | 84 | ||
| BC092212.1 | - | 4 - 5 | 383 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lonp1
Genomic Map