Assay Details
| Gene Symbol: Lonp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028782.2 | NP_083058.2 | 8 - 9 | 1368 | 101 | |
| GenBank mRNA | AK004820.1 | - | 8 - 9 | 1352 | 101 | |
| AK157474.1 | - | 8 - 9 | 1347 | 101 | ||
| AK160071.1 | - | 8 - 9 | 1353 | 101 | ||
| AK160076.1 | - | 8 - 9 | 1354 | 101 | ||
| AK161983.1 | - | 8 - 9 | 1353 | 101 | ||
| AK203362.1 | - | 1 - 2 | 54 | 101 | ||
| AY190302.1 | - | 8 - 9 | 1338 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lonp1
Genomic Map