Assay Details
| Gene Symbol: Gtf2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133801.2 | NP_598562.1 | 4 - 5 | 495 | 66 | |
| GenBank mRNA | AK013002.1 | - | 4 - 5 | 509 | 66 | |
| AK078048.1 | - | 4 - 5 | 508 | 66 | ||
| AK168236.1 | - | 4 - 5 | 508 | 66 | ||
| AK168243.1 | - | 4 - 5 | 508 | 66 | ||
| BC023081.1 | - | 4 - 5 | 495 | 5355475 | 66 | |
| BC031123.1 | - | 4 - 5 | 625 | 5026448 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2f1
Genomic Map