Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700022040.mm.1 |
NM_009451.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022040.mm.1 |
NM_009451.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022040.mm.1 |
TC1700002348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022041.mm.1 |
NM_009451.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022041.mm.1 |
NM_009451.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022041.mm.1 |
TC1700002348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022042.mm.1 |
NM_009451.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022042.mm.1 |
NM_009451.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022042.mm.1 |
TC1700002348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022044.mm.1 |
NM_009451.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022044.mm.1 |
NM_009451.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022044.mm.1 |
TC1700002348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022043.mm.1 |
NM_009451.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022043.mm.1 |
NM_009451.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022043.mm.1 |
TC1700002348.mm.1 |
MTA TC |