Assay Details
| Gene Symbol: Safb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029979.2 | NP_001025150.2 | 3 - 4 | 467 | 70 | |
| GenBank mRNA | AK147798.1 | - | 3 - 4 | 407 | 70 | |
| AK161772.1 | - | 3 - 4 | 447 | 70 | ||
| AK172898.1 | - | 3 - 4 | 443 | 70 | ||
| BC022609.1 | - | 3 - 4 | 408 | 70 | ||
| BC050855.1 | - | 3 - 4 | 401 | 70 | ||
| BC057039.1 | - | 3 - 4 | 396 | 70 | ||
| CN723671.1 | - | 3 - 4 | 469 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Safb2
Genomic Map