Assay Details
| Gene Symbol: Pdk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145630.2 | NP_663605.1 | 7 - 8 | 888 | 67 | |
| GenBank mRNA | AK075879.1 | - | 7 - 8 | 884 | 67 | |
| AK075985.1 | - | 7 - 8 | 944 | 67 | ||
| AK088478.1 | - | 7 - 8 | 969 | 67 | ||
| AK132024.1 | - | 7 - 8 | 912 | 67 | ||
| AK169874.1 | - | 7 - 8 | 969 | 67 | ||
| BC008126.1 | - | 7 - 8 | 888 | 3500763 | 67 | |
| CT010338.1 | - | 7 - 8 | 745 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdk3
Genomic Map