Assay Details
| Gene Symbol: Il18rap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010553.4 | NP_034683.1 | 10 - 11 | 1494 | 59 | |
| GenBank mRNA | AF077347.1 | - | 10 - 11 | 1503 | 59 | |
| BC120598.1 | - | 10 - 11 | 1501 | 40129521 | 59 | |
| BC120600.1 | - | 10 - 11 | 1501 | 40129523 | 59 | |
More Information
Set Membership: |
|

Il18rap
Genomic Map