Assay Details
Gene Symbol: Rab21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024454.1 | NP_077774.1 | 3 - 4 | 562 | 118 | |
GenBank mRNA | AB232615.1 | - | 3 - 4 | 317 | 118 | |
AK035980.1 | - | 2 - 3 | 131 | 118 | ||
AK172891.1 | - | 3 - 4 | 562 | 118 | ||
BC055042.1 | - | 3 - 4 | 562 | 6490069 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
