Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10600802 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10600806 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10600805 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10600801 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10600803 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10412037 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10600804 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10600797 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10412036 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0X00005753.mm.1 |
NM_001199337.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005753.mm.1 |
NM_001199337.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005753.mm.1 |
NM_001199338.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005753.mm.1 |
NM_001199338.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005753.mm.1 |
NM_001199339.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005753.mm.1 |
NM_001199339.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005753.mm.1 |
NM_026673.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005753.mm.1 |
NM_026673.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005753.mm.1 |
NR_004438.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005753.mm.1 |
TC0X00000897.mm.1 |
MTA TC |