Assay Details
| Gene Symbol: Sbspon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033288.3 | NP_001028460.1 | 3 - 4 | 568 | 84 | |
| GenBank mRNA | AK137144.1 | - | 3 - 4 | 569 | 84 | |
| AK143251.1 | - | 3 - 4 | 542 | 84 | ||
| BC147194.1 | - | 3 - 4 | 555 | 9056240 | 84 | |
| BC147195.1 | - | 3 - 4 | 555 | 9056224 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sbspon
Genomic Map