Assay Details
Gene Symbol: Etaa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026576.3 | NP_080852.2 | 1 - 2 | 320 | 105 | |
GenBank mRNA | AK019965.1 | - | 1 - 2 | 321 | 105 | |
AK034138.1 | - | 1 - 2 | 318 | 105 | ||
BC028916.1 | - | 1 - 2 | 315 | 3676092 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK034138.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
