Assay Details
| Gene Symbol: 4930519F16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029170.1 | NP_083446.1 | 2 - 3 | 259 | 104 | |
| GenBank mRNA | AK015842.1 | - | 2 - 3 | 104 | ||
| AK039711.1 | - | 2 - 3 | 259 | 104 | ||
| BC107248.2 | - | 2 - 3 | 261 | 40052342 | 104 | |
| BC107249.1 | - | 2 - 3 | 257 | 40052344 | 104 | |
| BC114984.1 | - | 2 - 3 | 261 | 40052347 | 104 | |
| BC115753.1 | - | 2 - 3 | 259 | 40052345 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

4930519F16Rik
Genomic Map