Assay Details
Gene Symbol: Nek11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172461.3 | NP_766049.2 | 7 - 8 | 1034 | 75 | |
GenBank mRNA | AK030042.1 | - | 7 - 8 | 993 | 75 | |
AK030186.1 | - | 6 - 7 | 910 | 75 | ||
AK043543.1 | - | 7 - 8 | 1273 | 75 | ||
AK054237.1 | - | 7 - 8 | 928 | 75 | ||
BC119588.1 | - | 6 - 7 | 868 | 40058889 | 75 | |
BC119589.1 | - | 6 - 7 | 868 | 40058890 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
