Assay Details
| Gene Symbol: Ryk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042607.1 | NP_001036072.1 | 9 - 10 | 1191 | 114 | |
| NM_001284258.1 | NP_001271187.1 | 8 - 9 | 1069 | 114 | ||
| NM_013649.3 | NP_038677.3 | 9 - 10 | 1200 | 114 | ||
| GenBank mRNA | AK076449.1 | - | 3 - 4 | 293 | 114 | |
| AK144355.1 | - | 8 - 9 | 1009 | 114 | ||
| BC006963.1 | - | 5 - 6 | 499 | 114 | ||
| BC032275.1 | - | 9 - 10 | 925 | 5370100 | 114 | |
| L02210.1 | - | 9 - 10 | 1175 | 114 | ||
| L21707.1 | - | 9 - 10 | 1209 | 114 | ||
| L38394.1 | - | 9 - 10 | 1123 | 114 | ||
| M98547.1 | - | 9 - 10 | 1164 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ryk
Genomic Map