Assay Details
| Gene Symbol: Ryk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042607.1 | NP_001036072.1 | 5 - 6 | 734 | 70 | |
| NM_001284258.1 | NP_001271187.1 | 4 - 5 | 612 | 70 | ||
| NM_013649.3 | NP_038677.3 | 5 - 6 | 734 | 70 | ||
| GenBank mRNA | AK144355.1 | - | 4 - 5 | 552 | 70 | |
| BC006963.1 | - | 1 - 2 | 33 | 70 | ||
| BC032275.1 | - | 5 - 6 | 468 | 5370100 | 70 | |
| L02210.1 | - | 5 - 6 | 709 | 70 | ||
| L21707.1 | - | 5 - 6 | 744 | 70 | ||
| L38394.1 | - | 5 - 6 | 657 | 70 | ||
| M98547.1 | - | 5 - 6 | 698 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ryk
Genomic Map