Assay Details
| Gene Symbol: Ryk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042607.1 | NP_001036072.1 | 6 - 7 | 878 | 77 | |
| NM_001284258.1 | NP_001271187.1 | 5 - 6 | 756 | 77 | ||
| NM_013649.3 | NP_038677.3 | 6 - 7 | 878 | 77 | ||
| GenBank mRNA | AK144355.1 | - | 5 - 6 | 696 | 77 | |
| BC006963.1 | - | 2 - 3 | 177 | 77 | ||
| BC032275.1 | - | 6 - 7 | 612 | 5370100 | 77 | |
| L02210.1 | - | 6 - 7 | 853 | 77 | ||
| L21707.1 | - | 6 - 7 | 888 | 77 | ||
| L38394.1 | - | 6 - 7 | 801 | 77 | ||
| M98547.1 | - | 6 - 7 | 842 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ryk
Genomic Map