Assay Details
| Gene Symbol: Vipr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009511.2 | NP_033537.1 | 11 - 12 | 1214 | 90 | |
| GenBank mRNA | BC138572.1 | - | 11 - 12 | 1112 | 8861592 | 90 |
| D28132.1 | - | 11 - 12 | 1146 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vipr2
Genomic Map