Assay Details
Gene Symbol: Gm4924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ459435.1 | - | 397 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062593 | DQ459435.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067112 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2067112 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07126828 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07126848 | - | - |
Whole Mouse Genome G4122A | A_52_P500597 | AK149107.1 | mouse_transcriptome |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27292195 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27292185 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08370171 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27292210 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08903564 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08903569 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08838349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27292193 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27292198 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08445356 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27292208 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27292200 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27292180 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08419316 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08276228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08276224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27292178 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27292191 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27292179 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08419317 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27292205 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27292183 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10365226 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10365223 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10365222 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10365224 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10365220 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10365221 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10365219 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10365225 | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 237412_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 237412_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1254729 | - | - |
