Assay Details
| Gene Symbol: Bambi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026505.2 | NP_080781.1 | 1 - 2 | 327 | 99 | |
| GenBank mRNA | AK011305.1 | - | 1 - 2 | 350 | 99 | |
| AK017303.1 | - | 1 - 2 | 329 | 99 | ||
| AK045192.1 | - | 1 - 2 | 327 | 99 | ||
| AK143185.1 | - | 1 - 2 | 341 | 99 | ||
| AK164632.1 | - | 1 - 2 | 330 | 99 | ||
| BC019378.1 | - | 1 - 2 | 309 | 5009872 | 99 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bambi
Genomic Map