Assay Details
| Gene Symbol: Smap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290683.1 | NP_001277612.1 | 3 - 4 | 288 | 198 | |
| NM_028534.3 | NP_082810.1 | 3 - 4 | 558 | 198 | ||
| NR_110972.1 | 3 - 4 | 288 | 194 | |||
| GenBank mRNA | AK014888.1 | - | 3 - 4 | 242 | 200 | |
| AK014966.1 | - | 3 - 4 | 242 | 198 | ||
| AK132868.1 | - | 3 - 4 | 289 | 194 | ||
| BC006946.1 | - | 3 - 4 | 466 | 2810817 | 198 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smap1
Genomic Map