Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900009085.mm.1 |
NM_001003911.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009085.mm.1 |
NM_001003911.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009085.mm.1 |
TC0900001158.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009087.mm.1 |
NM_001003911.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009087.mm.1 |
NM_001003911.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009087.mm.1 |
TC0900001158.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009089.mm.1 |
NM_001003911.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009089.mm.1 |
NM_001003911.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009089.mm.1 |
TC0900001158.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009088.mm.1 |
NM_001003911.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009088.mm.1 |
NM_001003911.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009088.mm.1 |
TC0900001158.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009092.mm.1 |
NM_001003911.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009092.mm.1 |
NM_001003911.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009092.mm.1 |
TC0900001158.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009090.mm.1 |
NM_001003911.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009090.mm.1 |
NM_001003911.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009090.mm.1 |
TC0900001158.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009091.mm.1 |
NM_001003911.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009091.mm.1 |
NM_001003911.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009091.mm.1 |
TC0900001158.mm.1 |
MTA TC |