Assay Details
| Gene Symbol: Tmed3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025360.2 | NP_079636.2 | 1 - 2 | 298 | 69 | |
| GenBank mRNA | AK002698.1 | - | 1 - 2 | 246 | 69 | |
| AK004545.1 | - | 1 - 2 | 246 | 69 | ||
| AK007544.1 | - | 1 - 2 | 240 | 69 | ||
| BC023338.1 | - | 1 - 2 | 192 | 5253777 | 69 | |
| BY159164.1 | - | 1 - 2 | 298 | 69 | ||
More Information
Set Membership: |
|

Tmed3
Genomic Map