Assay Details
Gene Symbol: Top3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009410.2 | NP_033436.1 | 13 - 14 | 1864 | 65 | |
GenBank mRNA | AB006074.1 | - | 13 - 14 | 1865 | 65 | |
AK029987.1 | - | 10 - 11 | 1257 | 65 | ||
AK037640.1 | - | 13 - 14 | 1765 | 65 | ||
AK164443.1 | - | 13 - 14 | 1765 | 65 | ||
AK170291.1 | - | 14 - 15 | 1671 | 65 | ||
AK171286.1 | - | 14 - 15 | 1671 | 65 | ||
BC099689.1 | - | 13 - 14 | 1679 | 30731611 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
