Assay Details
Gene Symbol: Dpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133803.2 | NP_598564.2 | 15 - 16 | 1767 | 72 | |
GenBank mRNA | AK031020.1 | - | 15 - 16 | 1732 | 72 | |
AK039975.1 | - | 11 - 12 | 3199 | 72 | ||
AK041330.1 | - | 2 - 3 | 436 | 72 | ||
AK087994.1 | - | 15 - 16 | 1731 | 72 | ||
AK149853.1 | - | 15 - 16 | 1732 | 72 | ||
AK150101.1 | - | 15 - 16 | 1727 | 72 | ||
AK150126.1 | - | 15 - 16 | 1721 | 72 | ||
AK166842.1 | - | 15 - 16 | 1732 | 72 | ||
AK168195.1 | - | 15 - 16 | 1720 | 72 | ||
BC004600.1 | - | 14 - 15 | 1709 | 3584924 | 72 | |
BC066817.1 | - | 15 - 16 | 1718 | 30429618 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
