Assay Details
| Gene Symbol: Kdm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001984.2 | NP_001001984.2 | 4 - 5 | 1088 | 91 | |
| GenBank mRNA | AK037157.1 | - | 5 - 6 | 985 | 91 | |
| AK147028.1 | - | 4 - 5 | 1080 | 91 | ||
| AK155866.1 | - | 4 - 5 | 1093 | 91 | ||
| BC076576.1 | - | 4 - 5 | 654 | 6840591 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdm2a
Genomic Map