Assay Details
Gene Symbol: Ssh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198113.2 | NP_932781.1 | 5 - 6 | 618 | 67 | |
GenBank mRNA | AB099289.1 | - | 5 - 6 | 616 | 67 | |
AK038981.1 | - | 4 - 5 | 1190 | 67 | ||
AK148293.1 | - | 5 - 6 | 611 | 67 | ||
AK149880.1 | - | 5 - 6 | 627 | 67 | ||
AK164662.1 | - | 4 - 5 | 316 | 67 | ||
AK166342.1 | - | 5 - 6 | 624 | 67 | ||
BC028922.1 | - | 5 - 6 | 618 | 3987714 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
