Assay Details
| Gene Symbol: Uprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081189.1 | NP_001074658.1 | 5 - 6 | 911 | 139 | |
| GenBank mRNA | BC147845.1 | - | 5 - 6 | 769 | 9087562 | 139 |
| BC147853.1 | - | 5 - 6 | 769 | 9087570 | 139 | |
More Information
Set Membership: |
|

Uprt
Genomic Map