Assay Details
Gene Symbol: Spast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162870.1 | NP_001156342.1 | 15 - 16 | 1737 | 130 | |
NM_016962.2 | NP_058658.2 | 15 - 16 | 1734 | 130 | ||
GenBank mRNA | AJ246002.1 | - | 15 - 16 | 1359 | 130 | |
AK007793.1 | - | 15 - 16 | 1514 | 130 | ||
AK129282.1 | - | 15 - 16 | 1711 | 130 | ||
BC046286.1 | - | 15 - 16 | 1734 | 6441742 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
