Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700010877.mm.1 |
NM_001162870.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010877.mm.1 |
NM_001162870.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010877.mm.1 |
NM_016962.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010877.mm.1 |
NM_016962.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010877.mm.1 |
TC1700001156.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010875.mm.1 |
NM_001162870.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010875.mm.1 |
NM_001162870.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010875.mm.1 |
NM_016962.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010875.mm.1 |
NM_016962.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010875.mm.1 |
TC1700001156.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010876.mm.1 |
NM_001162870.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010876.mm.1 |
NM_001162870.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010876.mm.1 |
NM_016962.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010876.mm.1 |
NM_016962.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010876.mm.1 |
TC1700001156.mm.1 |
MTA TC |