Assay Details
Gene Symbol: Mybl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290397.1 | NP_001277326.1 | 4 - 5 | 545 | 128 | |
NM_008651.3 | NP_032677.2 | 4 - 5 | 545 | 128 | ||
GenBank mRNA | BC101945.1 | - | 4 - 5 | 289 | 128 | |
BC103533.1 | - | 4 - 5 | 290 | 40039678 | 128 | |
BC103534.1 | - | 4 - 5 | 289 | 128 | ||
L35261.1 | - | 4 - 5 | 544 | 128 | ||
X82327.1 | - | 4 - 5 | 290 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
