Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700022478.mm.1 |
NM_001278415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022478.mm.1 |
NM_028388.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022478.mm.1 |
TC1700002422.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022479.mm.1 |
NM_001278415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022479.mm.1 |
NM_028388.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022479.mm.1 |
TC1700002422.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022480.mm.1 |
NM_001278415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022480.mm.1 |
NM_028388.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022480.mm.1 |
TC1700002422.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022486.mm.1 |
NM_001278415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022486.mm.1 |
NM_028388.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022486.mm.1 |
TC1700002422.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022490.mm.1 |
NM_001278415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022490.mm.1 |
NM_028388.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022490.mm.1 |
TC1700002422.mm.1 |
MTA TC |