Assay Details
| Gene Symbol: Ndufv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278415.1 | NP_001265344.1 | 5 - 6 | 489 | 123 | |
| NM_028388.3 | NP_082664.1 | 4 - 5 | 392 | 123 | ||
| GenBank mRNA | AK013511.1 | - | 4 - 5 | 343 | 123 | |
| AK078351.1 | - | 4 - 5 | 336 | 123 | ||
| AK088193.1 | - | 1 - 2 | 131 | 123 | ||
| AK146998.1 | - | 4 - 5 | 326 | 123 | ||
| AK150404.1 | - | 5 - 6 | 426 | 123 | ||
| AK151729.1 | - | 5 - 6 | 426 | 123 | ||
| AK159460.1 | - | 4 - 5 | 309 | 123 | ||
| AK166539.1 | - | 4 - 5 | 353 | 123 | ||
| AK169143.1 | - | 4 - 5 | 363 | 123 | ||
| BC030946.1 | - | 4 - 5 | 321 | 5003524 | 123 | |
| CF106105.1 | - | 3 - 4 | 234 | 123 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK150404.1, AK151729.1, NM_001278415.1 transcripts.
Set Membership: |
|

Ndufv2
Genomic Map