Assay Details
Gene Symbol: Themis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028998.1 | NP_083274.1 | 3 - 4 | 786 | 87 | |
GenBank mRNA | AK018663.1 | - | 3 - 4 | 787 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P118400 | NM_028998.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P118400 | AK018663.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P118400 | NM_028998.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P118400 | AK018663.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174743 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173615 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P118400 | NM_028998.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P118400 | AK018663.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P118400 | NM_028998.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P118400 | AK018663.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P118400 | NM_028998.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P118400 | AK018663.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P118400 | NM_028998.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P118400 | AK018663.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137529 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13200768 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306940 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307085 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306887 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307127 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306872 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306987 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307073 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306874 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306988 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306992 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13359665 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306956 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307053 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306980 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306938 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306958 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13487084 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306939 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306993 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307113 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306986 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306868 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307064 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307069 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307079 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307092 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307129 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13245102 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306942 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307067 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307088 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306954 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13308723 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306864 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307078 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307039 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307124 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306882 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13539421 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307100 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306880 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307072 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306945 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307062 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307132 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307058 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13247109 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307130 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13077427 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307095 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307110 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307099 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307104 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13084569 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306866 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13200765 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306861 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306937 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13197443 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307071 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307098 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306875 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13084568 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307048 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307050 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13358981 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307045 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307041 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307054 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307081 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306873 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307068 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306961 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306888 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306950 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306983 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13247101 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13337119 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13203784 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307070 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307105 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306863 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306877 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306990 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13200779 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307131 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307123 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307044 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307119 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13358979 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306869 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306944 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306957 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307059 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307122 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306982 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306860 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307093 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307087 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306870 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307066 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307084 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306878 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306952 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307042 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31307076 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306978 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31306946 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13539418 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1458210_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10452566 | - | - |
Mouse Genome 430 2.0 Array | 1458210_at | - | - |
Mouse Genome 430 2.0 Array | 1458210_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74556_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74556_PM_at | - | - |
Mouse Expression Set 430 | 1458210_at | - | - |
Mouse Expression Set 430 | 1458210_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1458210_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1458210_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 801583 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 106040022 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1225257 | - | - |
