Assay Details
| Gene Symbol: Gm1123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080776.1 | NP_001074245.1 | 7 - 8 | 1238 | 118 | |
| GenBank mRNA | AB262662.1 | - | 7 - 8 | 1267 | 118 | |
| BC114986.1 | - | 7 - 8 | 1238 | 40056015 | 118 | |
| BC115956.1 | - | 7 - 8 | 1238 | 40056016 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2172624 | BC114986.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2172624 | BC115956.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2172624 | NM_001080776.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2172624 | AB262662.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334800 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672829 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 831755 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 88916 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125445 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 784278 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809486 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 710534 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411582 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729046 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915013 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456007 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 700388 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561676 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 52421 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 455512 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 707287 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 691889 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852286 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127274 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 23786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156830 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 82648 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105781 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2172624 | BC114986.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2172624 | BC115956.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2172624 | NM_001080776.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2172624 | AB262662.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02009819 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06896825 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06896808 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06896864 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105781 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138172 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02009819 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26761902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07848793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08169109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07848805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07496250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07598146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07657971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07833664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07848790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26761933 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10595918 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10595920 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10595922 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10595923 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10595921 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10595915 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10595916 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10595919 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10595917 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457269_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10595914 | - | - |
| Mouse Genome 430 2.0 Array | 1457269_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457269_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 382097_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 382097_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900023990.mm.1 | NM_001080776.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900023990.mm.1 | NM_001080776.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900023990.mm.1 | TC0900002914.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1457269_PM_at | - | - |
| Mouse Expression Set 430 | 1457269_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457269_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457269_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 379273 | - | - |
| ABI Mouse Genome Survey Microarray | 670026 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239800 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2865047 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2865047 | - | - |

Gm1123
Genomic Map