Assay Details
| Gene Symbol: Mrps22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025485.3 | NP_079761.1 | 4 - 5 | 676 | 77 | |
| GenBank mRNA | AK003836.1 | - | 3 - 4 | 444 | 77 | |
| AK013925.1 | - | 4 - 5 | 657 | 77 | ||
| AK154600.1 | - | 4 - 5 | 657 | 77 | ||
| BC051198.1 | - | 4 - 5 | 656 | 1396237 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrps22
Genomic Map