Assay Details
| Gene Symbol: Copb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015827.2 | NP_056642.1 | 13 - 14 | 1634 | 76 | |
| GenBank mRNA | AF043120.1 | - | 2 - 3 | 179 | 76 | |
| AK088064.1 | - | 13 - 14 | 1635 | 76 | ||
| AK153352.1 | - | 13 - 14 | 1635 | 76 | ||
| BC006675.1 | - | 13 - 14 | 1621 | 3499585 | 76 | |
| BC010842.1 | - | 13 - 14 | 1623 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Copb2
Genomic Map